Skip to Content
Merck
All Photos(1)

Key Documents

931640

Sigma-Aldrich

Fmoc-L-W7N-OH

Novabiochem®, ≥95%

Synonym(s):

(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(1H-pyrrolo[2,3-b]pyridin-3-yl)propanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H21N3O4
CAS Number:
Molecular Weight:
427.45
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.21
form:
powder
Assay:
≥95%

Quality Level

Assay

≥95%

form

powder

mp

229-234 °C

storage temp.

−20°C

SMILES string

[nH]1c2ncccc2c(c1)C[C@H](NC(=O)OCC3c4c(cccc4)c5c3cccc5)C(=O)O

InChI

1S/C25H21N3O4/c29-24(30)22(12-15-13-27-23-16(15)10-5-11-26-23)28-25(31)32-14-21-19-8-3-1-6-17(19)18-7-2-4-9-20(18)21/h1-11,13,21-22H,12,14H2,(H,26,27)(H,28,31)(H,29,30)/t22-/m0/s1

InChI key

SEWQKWXKJVYKOO-QFIPXVFZSA-N

Application

Fmoc-L-W7N-OH is an arginine analog that can be used to prepare novel peptides for usage as putative therapeutics or in the elucidation of structure-activity relationships.

Legal Information

Product of Peptidream
Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service