917257
Empty cartridges
3 mL, amber, 50/box
Synonym(s):
3d bioprinting consumables, Syringe barrels
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121800
Recommended Products
Quality Level
description
3mL cartridges (50/box), amber, for UV and light-sensitive materials (up to 550 nm)
diam. × L
11.1 mm (0.44′′) × 73.0 mm (2.88′′)
Application
Empty cartridges for 3D Bioprinting application. The precision fit between the barrel and piston ensures consistent fluid deposition. The wiper piston improves fluid control and prevents fluid from dripping. Pistons, end, and tip caps are sold separately.
Transparent amber cartridges for UV and light-sensitive materials (up to 550 nm)
Transparent amber cartridges for UV and light-sensitive materials (up to 550 nm)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service