Skip to Content
Merck
All Photos(1)

Key Documents

794503

Sigma-Aldrich

Ritter Trifluoroiodomethane-DMSO Reagent

Synonym(s):

2DMSO-CF3I, DMSO-trifluoromethyl iodide adduct

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H12F3IO2S2
Molecular Weight:
352.18
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

reaction suitability

reaction type: C-C Bond Formation

refractive index

n/D 1.466

density

1.529 at 25 °C

functional group

fluoro
iodo
sulfoxide

shipped in

dry ice

storage temp.

2-8°C

SMILES string

FC(F)(F)I.CS(C)=O.CS(C)=O

InChI

1S/2C2H6OS.CF3I/c2*1-4(2)3;2-1(3,4)5/h2*1-2H3;

InChI key

MMNWUCHAIFIKKV-UHFFFAOYSA-N

Application

Product is a more convenient, liquified version of trifluoroiodomethane, which can be utilized in photocatalytic and electrophilic pentafluoroethylation of olefins and heteroatoms respectively.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

214.0 °F

Flash Point(C)

101.11 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service