Skip to Content
Merck
All Photos(2)

Key Documents

697303

Sigma-Aldrich

Tris(tert-pentoxy)silanol

packaged for use in deposition systems

Synonym(s):

TPS

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2C(CH3)2O)3SiOH
CAS Number:
Molecular Weight:
306.51
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

≥99.0% (GC)
≥99.99% (trace metals analysis)

form

liquid

bp

96-99 °C/2-3 mmHg (lit.)

density

0.944 g/mL at 25 °C (lit.)

SMILES string

CCC(C)(C)O[Si](O)(OC(C)(C)CC)OC(C)(C)CC

InChI

1S/C15H34O4Si/c1-10-13(4,5)17-20(16,18-14(6,7)11-2)19-15(8,9)12-3/h16H,10-12H2,1-9H3

InChI key

ORJFXWYTRPGGRK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Atomic number of base material: 14 Silicon

Hazard Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

222.8 °F - closed cup

Flash Point(C)

106 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alfredo Mirandola et al.
Medical physics, 42(9), 5287-5300 (2015-09-04)
To describe the dosimetric commissioning and quality assurance (QA) of the actively scanned proton and carbon ion beams at the Italian National Center for Oncological Hadrontherapy. The laterally integrated depth-dose-distributions (IDDs) were acquired with the PTW Peakfinder, a variable depth
G Magro et al.
Physics in medicine and biology, 60(17), 6865-6880 (2015-08-25)
This study was performed to evaluate the accuracy of a commercial treatment planning system (TPS), in optimising proton pencil beam dose distributions for small targets of different sizes (5-30 mm side) located at increasing depths in water. The TPS analytical
Meng Wu et al.
Journal of the Optical Society of America. A, Optics, image science, and vision, 32(9), 1707-1716 (2015-09-15)
Considering no previous literature reveals the effectiveness of image similarity coherent with corresponding illuminant in color constancy, we propose an edge-moment-based algorithm using regularized regression in an illumination-coherent space in a divide-and-conquer way. To represent the scene images, we adopt
Jasper L T Heerkens et al.
Poultry science, 94(9), 2008-2017 (2015-07-19)
Feather pecking and high mortality levels are significant welfare problems in non-cage housing systems for laying hens. The aim of this study was to identify husbandry-related risk factors for feather damage, mortality, and egg laying performance in laying hens housed
Hendrik Thoen et al.
Radiotherapy and oncology : journal of the European Society for Therapeutic Radiology and Oncology, 115(1), 16-21 (2015-03-31)
Neoadjuvant chemoradiotherapy is used prior to surgery in curative treatment of esophageal cancer (EC). We evaluated the in-field control of this multimodal treatment by extraction of radiation dose parameters and determination of the spatial relation between tumor recurrence location(s) and

Articles

igma-Aldrich.com presents an article regarding the savannah ALD system - an excellent tool for atomic layer deposition.

High Purity Metalorganic Precursors for CPV Device Fabrication

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service