456624
Polyethylene-graft-maleic anhydride
viscosity 500 cP (140 °C)(lit.)
Synonym(s):
maleated copolymer, maleated polymer, maleic copolymer
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
composition
maleic anhydride, ~0.5 wt. %
viscosity
500 cP(140 °C)(lit.)
saponification value
6 mg KOH/g
transition temp
Tm (DSC) 107 °C (at peak)
solubility
H2O: insoluble
toluene and xylene: soluble
density
0.92 g/mL at 25 °C
InChI
1S/C4H2O3.C2H4/c5-3-1-2-4(6)7-3;1-2/h1-2H;1-2H2
InChI key
YYXLGGIKSIZHSF-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Bond aid for polar to nonpolar substances. Compatibilizer for polymer blends.
Features and Benefits
Contributes abrasion resistance, lubricity, antiblocking, and surface glass, allows chemical bonding via anhydride group.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Farmaco (Societa chimica italiana : 1989), 58(5), 391-396 (2003-05-06)
Bioadhesive tablet formulations have been developed for mucosal application. Sixteen different bioadhesive tablet formulations were prepared and evaluated. Their bioadhesion to vaginal mucosa were studied by tensile testing method. The swelling behaviour of the tablets in three different solutions was
Archives of toxicology, 87(1), 99-109 (2012-08-14)
To uncover the size influence of TiO(2) nanoparticles on their potential toxicity, the cytotoxicity of different-sized TiO(2) nanoparticles with and without photoactivation was tested. It was demonstrated that without photoactivation, TiO(2) nanoparticles were inert up to 100 μg/ml. On the contrary
Applied biochemistry and biotechnology, 11(3), 177-189 (1985-06-01)
Alkaline phosphatase, APase, (EC 3.1.31) from calf intestine, after shifting the equilibrium by effector molecules towards the dimeric form of the enzyme, was coupled (ratio 1:2, protein: copolymer) to a copolymer of ethylene and maleic anhydride, EMA. The water-soluble APase-EMA
Nucleosides, nucleotides & nucleic acids, 20(4-7), 895-899 (2001-09-21)
(Polymer-oligonucleotide) conjugates were obtained via direct ON synthesis from the poly(ethylene-alt-maleic anhydride: PEMA) grafted onto glass bead surfaces. The effect of the ON number per polymer chain on binding was evaluated through Tm experiments. Results were correlated with ELOSA (Enzyme
Journal of microencapsulation, 24(1), 11-19 (2007-04-19)
This paper shows two types of microcapsules used for electrophoretic display. One is prepared by in-situ polymerization which is based on urea, melamine and formaldehyde and another by complex coacervation, which is composed of gelatin and gum Arabic. Microcapsules attract
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service