About This Item
Recommended Products
isotopic purity
99 atom % D
Assay
≥99.00%
form
liquid
contains
1 % (v/v) TMS
expl. lim.
0.34-6.3 % (lit.)
technique(s)
NMR: suitable
impurities
≤0.0100% water
water
refractive index
n20/D 1.494 (lit.)
bp
110 °C (lit.)
mp
-84 °C (lit.)
density
0.943 g/mL at 25 °C (lit.)
mass shift
M+8
SMILES string
[2H]c1c([2H])c([2H])c(c([2H])c1[2H])C([2H])([2H])[2H]
InChI
1S/C7H8/c1-7-5-3-2-4-6-7/h2-6H,1H3/i1D3,2D,3D,4D,5D,6D
InChI key
YXFVVABEGXRONW-JGUCLWPXSA-N
Looking for similar products? Visit Product Comparison Guide
Recommended products
accessory
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3
Target Organs
Central nervous system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
39.2 °F
Flash Point(C)
4 °C
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service