HPLC Analysis of Urea, Carbamate, Triazine, Aniline Pesticides on Discovery® C18
Materials
Analytical column
Product No.
Description
Pricing
CONDITIONS
column
Discovery C18, 15 cm × 4.6 mm I.D., 5 μm particles (504955)
mobile phase
[A] acetonitrile; [B] water
flow rate
2 mL/min
column temp.
20 °C
detector
UV, 214 nm
injection
1 μL
Description
Legal Information
Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany