HPLC Analysis of Xylene Isomers on Discovery® Zr-Carbon
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Discovery Zr-Carbon, 15 cm × 4.6 mm I.D., 3 μm particles (65730-U)
mobile phase
[A] water: [B] acetonitrile; (50:50, A:B)
flow rate
1 mL/min
column temp.
30 °C
detector
UV, 254 nm
injection
1 μL
sample
o-xylene, m-xylene, p-xylene (290 μg/mL)
Description
Legal Information
Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany