Accéder au contenu
MilliporeSigma
HPLC Analysis of Synthetic Food Dyes on Ascentis® Express C18
HPLC Analysis of Synthetic Food Dyes on Ascentis® Express C18 application for HPLC
Matériaux
Colonne analytique
Réf. du produit
Description
Tarif
Colonne de CLHP Ascentis® Express C18, 2,7 μm
2.7 μm particle size, L × I.D. 10 cm × 2.1 mm
Étalon
Réf. du produit
Description
Tarif
Phloxine B
Dye content ≥80 %, certified by the BSC
Rose bengal
certified by the BSC, Dye content ≥80 %
Erioglaucine disodium salt
coloring dye
Tartrazine
analytical standard
Sunset Yellow FCF
analytical standard
Allura Red AC
analytical standard
Amaranth
analytical standard
Erythrosin B
analytical standard
Fast Green FCF
analytical standard
Indigo carmine
analytical standard
CONDITIONS
column
Ascentis Express C18, 10 cm x 2.1 mm I.D., 2.7 μm particles (53823-U)
mobile phase
[A] 100 mM ammonium phosphate monobasic, pH 7.0 with ammonium hydroxide; [B] water; [C] acetonitrile
gradient
10% A constant; 85 to 25% B, 5 to 65% C in 6 min; 25% B, 65% C held for 5 min
flow rate
0.3 mL/min
column temp.
40 °C
detector
UV, 254 nm
injection
5 μL
sample
20 μg/mL each in water
Description
Informations légales
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany