HPLC Analysis of Bisphenol A, Dimethylbisphenol A and Nonylphenol on Ascentis® Express RP-Amide (UV Detection)
![HPLC Analysis of Bisphenol A, Dimethylbisphenol A and Nonylphenol on Ascentis® Express RP-Amide (UV Detection) application for HPLC](/deepweb/assets/sigmaaldrich/product/images/387/638/fa36e360-2d46-45a0-a0f6-6a2500fce7d8/800/fa36e360-2d46-45a0-a0f6-6a2500fce7d8.jpg)
Matériaux
Colonne analytique
Réf. du produit
Description
Tarif
CONDITIONS
column
Ascentis Express RP-Amide, 5 cm x 4.6 mm I.D., 2.7 μm particles (53922-U)
mobile phase
[A] water; [B] acetonitrile
gradient
50 to 100% B in 10 min
flow rate
1.0 mL/min
column temp.
35 °C
detector
UV, 280 nm
injection
10 μL
sample
50 mg/L of each in 50:50, water: acetonitrile
Description
Remarque sur l'analyse
This application demonstrates the suitability of Ascentis Express C18 for the efficient separation of milk thistle related compounds
Informations légales
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany