Accéder au contenu
Merck
HPLC Analysis of Ascorbic Acid on Ascentis® RP-Amide
HPLC Analysis of Ascorbic Acid on Ascentis® RP-Amide application for HPLC
Matériaux
Colonne analytique
Réf. du produit
Description
Tarif
Colonne HPLC® Ascentis RP-Amide
5 μm particle size, L × I.D. 15 cm × 4.6 mm
CONDITIONS
column
Ascentis RP-Amide, 15 cm x 4.6 mm I.D., 5 μm particles (565324-U)
mobile phase
25 mM monobasic potassium phosphate (pH 3.0 with phosphoric acid)
flow rate
1 mL/min
column temp.
35 °C
detector
UV, 230 nm
injection
10 μL
sample
as indicated each in mobile phase
Description
Informations légales
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany