MilliporeSigma
HPLC Analysis of Acetaminophen, Caffeine, Chlorpheniramine, Ibuprofen, Naproxen, Phenylephrine, and Pseudoephedrine on Ascentis® Express C18
HPLC Analysis of Acetaminophen, Caffeine, Chlorpheniramine, Ibuprofen, Naproxen, Phenylephrine, and Pseudoephedrine on Ascentis® Express C18 application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Ascentis® Express C18, 2.7 μm HPLC Column
2.7 μm particle size, L × I.D. 5 cm × 2.1 mm
standard
Product No.
Description
Pricing
Caffeine
Sigma Reference Standard, vial of 250 mg
Acetaminophen
analytical standard
Maleic acid
ReagentPlus®, ≥99% (HPLC)
Naproxen
meets USP testing specifications
Ibuprofen
≥98% (GC)
(−)-Pseudoephedrine
purum, ≥96.0% (sum of enantiomers, GC)
Chlorpheniramine Maleate
Pharmaceutical Secondary Standard; Certified Reference Material
Phenylephrine
European Pharmacopoeia (EP) Reference Standard
CONDITIONS
column
Ascentis Express C18, 5 cm x 2.1 mm I.D., 2.7 μm particles (53822-U)
mobile phase
[A] 5 mM ammonium phosphate monobasic, pH 2.0 with phosphoric acid: acetonitrile (98:2); [B] acetonitrile;
gradient
0 to 68% B in 3 min; held at 68% B for 2 min
flow rate
0.4 mL/min
column temp.
35 °C
detector
UV, 210 nm
injection
1 μL
sample
100 μg/mL in 95:5, water: methanol
Description
Analysis Note
This application demonstrates the suitability of Ascentis Express C18 for the efficient separation of various over the counter drugs.
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany