MilliporeSigma
HPLC Analysis of Bovine Serum Albumin (BSA) on Zenix® SEC-300 versus SRT® SEC-300, Effect of Particle Size on Resolution
HPLC Analysis of Bovine Serum Albumin (BSA) on Zenix® SEC-300 versus SRT® SEC-300, Effect of Particle Size on Resolution application for HPLC
Materials
Analytical column
Product No.
Description
Pricing
Zenix® SEC-300 Gel Filtration Column, 3 μm
L × I.D. 30 cm × 7.8 mm, 3 μm particle size, stainless steel
SRT® SEC-300 Gel Filtration Column, 5 μm
L × I.D. 30 cm × 7.8 mm, 5 μm particle size, stainless steel
CONDITIONS
column
SRT SEC-300, 30 cm x 7.8 mm I.D., 5 μm, 300 Å particles (Z777051)
column
Zenix SEC-300, 30 cm x 7.8 mm I.D., 3 μm, 300 Å particles (Z777033)
mobile phase
150 mM sodium phosphate, pH 7
flow rate
1 mL/min
column temp.
ambient
detector
UV, 214 nm
injection
10 μL
sample
5 mg/mL, BSA
Description
Legal Information
SRT is a registered trademark of Sepax Technologies
Zenix is a registered trademark of Sepax Technologies