Merck
GC Analysis of Primary Amines on Carbowax® Amine
GC Analysis of Primary Amines on Carbowax® Amine suitable for GC
Materials
analytical column
Product No.
Description
Pricing
Carbowax® Amine Capillary GC Column
L × I.D. 30 m × 0.53 mm, df 1.00 μm
CONDITIONS
column
Carbowax Amine, 30m × 0.53 mm I.D., 1.0 μm (25353)
oven
50 °C, 8 °C/min to 200 °C (10 min)
inj. temp.
220 °C
detector
FID, 220 °C
carrier gas
helium, 20 cm/sec @ 170 °C
injection
0.02 μL 100:1 split
sample
neat amines mix
Description
Legal Information
Carbowax is a registered trademark of Union Carbide