Merck
GC Analysis of Geranium Bourbon Essential Oil on SUPELCOWAX 10
GC Analysis of Geranium Bourbon Essential Oil on SUPELCOWAX 10 suitable for GC
Materials
analytical column
Product No.
Description
Pricing
SUPELCOWAX 10 Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.25 μm
CONDITIONS
column
SUPELCOWAX 10, 30 m x 0.25 mm I.D., 0.25 μm (24079)
oven
50 °C (2 min), 2 °C/min to 230 °C (1 min)
detector
FID
carrier gas
helium, 20 cm/sec @ 155 °C
injection
0.2 μL, 100:1 split
sample
neat oil
Description
Legal Information
null