GC Analysis of Amino Acids (as TBDMS Derivatives) on SLB®-5ms (20 m x 0.18 mm I.D., 0.18 μm), Fast GC Analysis
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
SLB-5ms, 20 m × 0.18 mm I.D., 0.18 μm (28564-U)
oven
100 °C (1 min), 35 °C/min to 290 °C (3 min), 40 °C/min to 360 °C
inj. temp.
250 °C
MSD interface
325 °C
scan range
m/z = 40-450
carrier gas
helium, 1 mL/min, constant
injection
0.5 μL, splitless (1.0 min)
liner
2 mm I.D., straight
sample
TBDMS derivatives of amino acids, each approximately 23 ug/mL
Description
Legal Information
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany