>P04963ups3|PRXC_LEPFU Chloroperoxidase (Chain 21-319) - Leptoxyphium fumago (Caldariomyces fumago) QEPGSGIGYPYDNNTLPYVAPGPTDSRAPCPALNALANHGYIPHDGRAISRETLQNAFLNHMGIANSVIELALTNAFVVCEYVTGSDCGDSLVNLTLLAEPHAFEHDHSFSRKDYKQGVANSNDFIDNRNFDAETFQTSLDVVAGKTHFDYADMNEIRLQRESLSNELDFPGWFTESKPIQNVESGFIFALVSDFNLPDNDENPLVRIDWWKYWFTNESFPYHLGWHPPSPAREIEFVTSASSAVLAASVTSTPSSLPSGAIGPGAEAVPLSFASTMTPFLLATNAPYYAQDPTLGPND >P10547ups3|LSTP_STASI Lysostaphin (Chain 248-493) - Staphylococcus simulans AATHEHSAQWLNNYKKGYGYGPYPLGINGGMHYGVDFFMNIGTPVKAISSGKIVEAGWSNYGGGNQIGLIENDGVHRQWYMHLSKYNVKVGDYVKAGQIIGWSGSTGYSTAPHLHFQRMVNSFSNSTAQDPMPFLKSAGYGKAGGTVTPTPNTGWKTNKYGTLYKSESASFTPNTDIITRTTGPFRSMPQSGVLKAGQTIHYDEVMKQDGHVWVGYTGNSGQRIYLPVRTWNKSTNTLGVLWGTIK >A9X9K8ups3|A9X9K8_9LACT Pyruvate Oxidase (Chain 1-591) - Aerococcus viridans MSDNKINIGLAVMKILESWGADTIYGIPSGTLSSLMDAMGEEENNVKFLQVKHEEVGAMAAVMQSKFGGNLGVTVGSGGPGASHLINGLYDAAMDNIPVVAILGSRPQRELNMDAFQELNQNPMYDHIAVYNRRVAYAEQLPKLVDEAARMAIAKRGVAVLEVPGDFAKVEIDNDQWYSSANSLRKYEPIAPAAQDIDAAVELLNNSKRPVIYAGIGTMGHGPAVQELARKIKAPVITTGKNFETFEWDFEALTGSTYRVGWKPANETILEADTVLFAGSNFPFSEVEGTFRNVDNFIQIDIDPAMLGKRHHADVAILGDAGLAIDEILNKVDAVEESAWWTANLKNIANWREYINMLETKEEGDLQFYQVYNAINNHADEDAIYSIDVGNSTQTSIRHLHMTPKNMWRTSPLFATMGIAIPGGLGAKNTYPDRQVWNIIGDGAFSMTYPDVVTNVRYNMPVINVVFSNTEYAFIKNKYEDTNKNLFGVDFTDVDYAKIAEAQGAKGFTVSRIEDMDRVMAEAVAANKAGHTVVIDCKITQDRPIPVETLKLDSKLYSEDEIKAYKERYEAANLVPFREYLEAEGLESKYIK >P02866ups3|CONA_CANEN Lectin Concanavalin-A (Chain 164–281 + 30–148) - Canavalia ensiformis (Jack bean) (Dolichos ensiformis) ADTIVAVELDTYPNTDIGDPSYPHIGIDIKSVRSKKTAKWNMQNGKVGTAHIIYNSVDKRLSAVVSYPNADSATVSYDVDLDNVLPEWVRVGLSASTGLYKETNTILSWSFTSKLKSNSTHETNALHFMFNQFSKDQKDLILQGDATTGTDGNLELTRVSSNGSPQGSSVGRALFYAPVHIWESSAVVASFEATFTFLIKSPDSHPADGIAFFISNIDSSIPSGSTGRLLGLFPDAN >P00711ups3|LALBA_BOVIN Alpha-Lactalbumin bovine milk (Chain 20-142) - Bos taurus (Bovine) EQLTKCEVFRELKDLKGYGGVSLPEWVCTTFHTSGYDTQAIVQNNDSTEYGLFQINNKIWCKDDQNPHSSNICNISCDKFLDDDLTDDIMCVKKILDKVGINYWLAHKALCSEKLDQWLCEKL >P00698ups3|LYSC_CHICK Lysozyme C chicken egg white (Chain 19-147) - Gallus gallus (Chicken) KVFGRCELAAAMKRHGLDNYRGYSLGNWVCAAKFESNFNTQATNRNTDGSTDYGILQINSRWWCNDGRTPGSRNLCNIPCSALLSSDITASVNCAKKIVSDGNGMNAWVAWRNRCKGTDVQAWIRGCRL >P21163ups3|PNGF_ELIMR Peptide-N(4)-(N-acetyl-beta-D-glucosaminyl)asparagine amidase F (Chain 41-354) - Elizabethkingia miricola (Chryseobacterium miricola) APADNTVNIKTFDKVKNAFGDGLSQSAEGTFTFPADVTTVKTIKMFIKNECPNKTCDEWDRYANVYVKNKTTGEWYEIGRFITPYWVGTEKLPRGLEIDVTDFKSLLSGNTELKIYTETWLAKGREYSVDFDIVYGTPDYKYSAVVPVIQYNKSSIDGVPYGKAHTLGLKKNIQLPTNTEKAYLRTTISGWGHAKPYDAGSRGCAEWCFRTHTIAINNANTFQHQLGALGCSANPINNQSPGNWAPDRAGWCPGMAVPTRIDVLNNSLTGSTFSYEYKFQSWTNNGTNGDAFYAISSFVIAKSNTPISAPVVTN >P00921ups3|CAH2_BOVIN Carbonic Anhydrase 2 (Chain 2-260) - Bos taurus (Bovine) SHHWGYGKHNGPEHWHKDFPIANGERQSPVDIDTKAVVQDPALKPLALVYGEATSRRMVNNGHSFNVEYDDSQDKAVLKDGPLTGTYRLVQFHFHWGSSDDQGSEHTVDRKKYAAELHLVHWNTKYGDFGTAAQQPDGLAVVGVFLKVGDANPALQKVLDALDSIKTKGKSTDFPNFDPGSLLPNVLDYWTYPGSLTTPPLLESVTWIVLKEPISVSSQQMLKFRTLNFNAEGEPELLMLANWRPAQPLKNRQVRGFPK