Merck
mdm2 gene alterations and mdm2 protein expression in breast carcinomas.
The Journal of pathology (1995-01-01)
A Marchetti, F Buttitta, S Girlando, P Dalla Palma, S Pellegrini, P Fina, C Doglioni, G Bevilacqua, M Barbareschi
ABSTRACT
This study investigates the mdm2 gene status and expression in 66 surgically resected human breast carcinomas, with correlations with clinico-pathological and biological data (histological type, grading, steroid receptor status, p53 expression, proliferative activity). Four (7.7 per cent) out of 52 informative cases bear mdm2 gene amplification (four-to ten-fold) and 8 (15.4 per cent) of 52 cases showed borderline amplification (three-fold). Nine (13.6 per cent) out of 66 cases showed strong mdm2 nuclear immunoreactivity. Twenty-seven (40.9 per cent) cases showed isolated mdm2 reactive nuclei. All cases with clear amplification showed a high percentage of mdm2 immunoreactive nuclei. The relationship between gene amplification and mdm2 protein expression is highly significant (P < 0.0001). No association was observed between mdm2 gene amplification and any of the considered clinico-pathological and biological parameters, while mdm2 immunoreactivity showed a significant association only with oestrogen receptor immunoreactivity (P = 0.009). p53 expression was associated neither with mdm2 gene amplification nor with mdm2 immunoreactivity. It could be tempting to hypothesize that the evaluation of the combined mdm2/p53 immunohistochemical phenotype in human breast carcinoma could give us better prognostic information than the evaluation of the expression of the p53 protein alone.