Ammonium persulfate
Synonym(s):
AP, APS, Ammonium peroxodisulfate, Ammonium peroxydisulfate, PER
Linear Formula:
(NH4)2S2O8
CAS Number:
Molecular Weight:
228.20
Products
Product Category
Brand
Feature
Formula Weight
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Markush Class
Markush Group
Reagent Type
Special Grade
Technique
Showing 1-9 of 9
Compare
Product Number
Description
Pricing