Merck
Gender difference in kidney electrolyte transport. I. Role of AT
American journal of physiology. Renal physiology (2017-06-02)
Jing Li, Ryo Hatano, Shuhua Xu, Laxiang Wan, Lei Yang, Alan M Weinstein, Lawrence Palmer, Tong Wang
ABSTRACT
We studied gender differences in Na
Sigma-Aldrich
Inactin® hydrate, ≥98% (HPLC)