Merck
Liraglutide reduces body weight by upregulation of adenylate cyclase 3.
Nutrition & diabetes (2017-05-10)
Z Li, Y Liang, N Xia, Y Lai, H Pan, S Zhou, F Jiang, Y He
ABSTRACT
According to recent studies, adenylate cyclase 3 (AC3) is associated with obesity. Liraglutide reduces blood glucose levels and body weight (BW). We performed a 2 × 2 factorial experiment to study the relationships among AC3, liraglutide and obesity and to obtain a more comprehensive understanding of the mechanisms underlying the physiological effects of liraglutide on obesity. A high-fat diet was used to induce obesity in C57BL/6J mice. Both the normal and obese mice were treated with liraglutide (1 mg kg The administration of liraglutide significantly decreased the BW in obese mice and normal control mice. The BW of obese mice exhibited a more obvious decrease. Hepatic AC3 mRNA and protein levels and serum AC3 levels were significantly reduced in obese mice compared with those in normal control mice. The administration of liraglutide significantly increased the hepatic expression of the AC3 and GLP-1R mRNAs and proteins and serum AC3 levels. The hepatic expression of the AC3 mRNA and protein and serum AC3 levels were negatively correlated with BW loss in the liraglutide-treated group. Pearson's correlation coefficients for these comparisons are r=-0.448, P=0.048; r=-0.478, P=0.046; and r=-0.909, P=0.000, respectively. Based on our research, liraglutide reduces BW, possibly by increasing the expression of AC3.