Merck
  • In situ proliferation contributes to accumulation of tumor-associated macrophages in spontaneous mammary tumors.
In situ proliferation contributes to accumulation of tumor-associated macrophages in spontaneous mammary tumors.
European journal of immunology (2014-05-07)
Piotr Tymoszuk, Hanneke Evens, Vanessa Marzola, Katarzyna Wachowicz, Marie-Helene Wasmer, Sebak Datta, Elisabeth Müller-Holzner, Heidi Fiegl, Günther Böck, Nico van Rooijen, Igor Theurl, Wolfgang Doppler
ABSTRACT
Infiltration of a neoplasm with tumor-associated macrophages (TAMs) is considered an important negative prognostic factor and is functionally associated with tumor vascularization, accelerated growth, and dissemination. However, the ontogeny and differentiation pathways of TAMs are only incompletely characterized. Here, we report that intense local proliferation of fully differentiated macrophages rather than low-pace recruitment of blood-borne precursors drives TAM accumulation in a mouse model of spontaneous mammary carcinogenesis, the MMTVneu strain. TAM differentiation and expansion is regulated by CSF1, whose expression is directly controlled by STAT1 at the gene promoter level. These findings appear to be also relevant for human breast cancer, in which an interrelationship between STAT1, CSF1, and macrophage marker expression was identified. We propose that, akin to various MU subtypes in nonmalignant tissues, local proliferation and CSF1 play a vital role in the homeostasis of TAMs.