Merck
  • Matrix metalloproteinase-7 and matrix metalloproteinase-25 in oral tongue squamous cell carcinoma.
Matrix metalloproteinase-7 and matrix metalloproteinase-25 in oral tongue squamous cell carcinoma.
Head & neck (2014-02-04)
Laura K Mäkinen, Valtteri Häyry, Jaana Hagström, Timo Sorsa, Fabricio Passador-Santos, Harri Keski-Säntti, Jari Haukka, Antti A Mäkitie, Caj Haglund, Timo Atula
ABSTRACT
Predicting the clinical course of early-stage oral tongue squamous cell carcinoma (SCC) is challenging. As matrix metalloproteinases (MMPs) are enzymes associated with invasion, metastasis, and poor survival in many cancers, we examined MMP-7 and MMP-25 in oral tongue SCC. We used tissue microarray (TMA) technique and immunohistochemistry to study the expression of MMP-7 and MMP-25 in 73 patients with stage I to II oral tongue SCC and compared their immunoexpressions with clinical data. Immunohistochemistry revealed MMP-7 and MMP-25 expression in 90% (n = 63 of 70) and 90% (n = 64 of 71) of the tumors, respectively. MMP-7 protein expression was associated with presence of occult cervical metastases (odds ratio [OR], 3.67; p = .013), increased invasion depth (OR, 4.60; p = .005), and higher tumor grade (OR, 3.30; p = .007). MMP-7 expression was predictive for poor outcome (p = .021). Immunostaining of MMP-25 did not correlate with any clinical parameters. We conclude that MMP-7, but not MMP-25, expression may have prognostic significance in early-stage oral tongue SCC.