Merck
  • Muscarinic receptor subtypes involved in regulation of colonic motility in mice: functional studies using muscarinic receptor-deficient mice.
Muscarinic receptor subtypes involved in regulation of colonic motility in mice: functional studies using muscarinic receptor-deficient mice.
European journal of pharmacology (2011-09-20)
Takaji Kondo, Miwa Nakajima, Hiroki Teraoka, Toshihiro Unno, Sei-ichi Komori, Masahisa Yamada, Takio Kitazawa
ABSTRACT
Although muscarinic M(2) and M(3) receptors are known to be important for regulation of gastric and small intestinal motility, muscarinic receptor subtypes regulating colonic function remain to be investigated. The aim of this study was to characterize muscarinic receptors involved in regulation of colonic contractility. M(2) and/or M(3) receptor knockout (KO) and wild-type mice were used in in vivo (defecation, colonic propulsion) and in vitro (contraction) experiments. Amount of feces was significantly decreased in M(3)R-KO and M(2)/M(3)R-KO mice but not in M(2)R-KO mice. Ranking of colonic propulsion was wild-type=M(2)R-KO>M(3)R-KO>M(2)/M(3)R-KO. In vitro, the amplitude of migrating motor complexes in M(2)R-KO, M(3)R-KO and M(2)/M(3)R-KO mice was significantly lower than that in wild-type mice. Carbachol caused concentration-dependent contraction of the proximal colon and distal colon from wild-type mice. In M(2)R-KO mice, the concentration-contraction curves shifted to the right and downward. In contrast, carbachol caused non-sustained contraction and relaxation in M(3)R-KO mice depending on its concentration. Carbachol did not cause contraction but instead caused relaxation of colonic strips from M(2)/M(3)R-KO mice. 4-[[[(3-chlorophenyl)amino]carbonyl]oxy]-N,N,N-trimethyl-2-butyn-1-aminium chloride (McN-A-343) caused a non-sustained contraction of colonic strips from wild-type mice, and this contraction was changed to a sustained contraction by tetrodotoxin, pirenzepine and L-nitroarginine methylester (L-NAME). In the colon of M(2)/M(3)R-KO mice, McN-A-343 caused only relaxation, which was decreased by tetrodotoxin, pirenzepine and L-NAME. In conclusion, M(1), M(2) and M(3) receptors regulate colonic motility of the mouse. M(2) and M(3) receptors mediate cholinergic contraction, but M(1) receptors on inhibitory nitrergic nerves counteract muscarinic contraction.
Sigma-Aldrich
McN-A-343, ≥98% (TLC)