Merck
  • Myeloid HIF-1α regulates pulmonary inflammation during experimental Mycobacterium tuberculosis infection.
Myeloid HIF-1α regulates pulmonary inflammation during experimental Mycobacterium tuberculosis infection.
Immunology (2019-10-14)
Mariana Resende, Catarina M Ferreira, Ana Margarida Barbosa, Marcos S Cardoso, Jeremy Sousa, Margarida Saraiva, António G Castro, Rui Appelberg, Egídio Torrado
ABSTRACT
The transcription factor hypoxia-inducible factor-1 alpha (HIF-1α) is a key regulator of the response and function of myeloid cells in hypoxic and inflammatory microenvironments. To define the role of HIF-1α in tuberculosis, the progression of aerosol Mycobacterium tuberculosis infection was analysed in mice deficient in HIF-1α in the myeloid lineage (mHIF-1α-/- ). We show that myeloid HIF-1α is not required for the containment of the infection, as both wild-type (WT) and mHIF-1α-/- mice mounted normal Th1 responses and maintained control of bacterial growth throughout infection. However, during chronic infection mHIF-1α-/- mice developed extensive lymphocytic inflammatory involvement of the interstitial lung tissue and died earlier than WT mice. These data support the hypothesis that HIF-1α activity coordinates the response of myeloid cells during M. tuberculosis infection to prevent excessive leucocyte recruitment and immunopathological consequences to the host.