Merck
  • A flexible approach to azasugars: asymmetric total syntheses of (+)-castanospermine, (+)-7-deoxy-6-epi-castanospermine, and (+)-1-epi-castanospermine.
A flexible approach to azasugars: asymmetric total syntheses of (+)-castanospermine, (+)-7-deoxy-6-epi-castanospermine, and (+)-1-epi-castanospermine.
Chemistry (Weinheim an der Bergstrasse, Germany) (2010-04-10)
Gang Liu, Tian-Jun Wu, Yuan-Ping Ruan, Pei-Qiang Huang
ABSTRACT
The asymmetric total synthesis of natural azasugars (+)-castanospermine, (+)-7-deoxy-6-epi-castanospermine, and synthetic (+)-1-epi-castanospermine has been accomplished in nine to ten steps from a common chiral building block (S)-8. The method features a powerful chiral relay strategy consisting of a highly diastereoselective vinylogous Mukaiyama-type reaction with either chiral or achiral aldehydes (> or = 95% de; de=diastereomeric excess) and a diastereodivergent reduction of tetramic acids, which allows formation of three continuous stereogenic centers with high diastereoselectivities. The method also provides a flexible access to structural arrays of 5-(alpha-hydroxyalkyl)tetramic acids, such as 17/34, and 5-(alpha-hydroxyalkyl)-4-hydroxyl-2-pyrrolidinones, such as 18 and 25/35 a. The method constitutes the first realization of the challenging chiral synthons A and D and thus of the conceptually attractive retrosynthetic analysis shown in Scheme 1 in a highly enantioselective manner.
Sigma-Aldrich
(1S,6S,7R,8R,8aR)-1,6,7,8-Tetrahydroxyindolizidine, 98%
Sigma-Aldrich
Castanospermine, ≥94% (GC), BioUltra, from Castanospermum australe seeds