Triadimenol A
Synonym(s):
(αS,βR)-rel-α-tert-Butyl-β-(4-chlorophenoxy)-1H-1,2,4-triazole-1-ethanol, (αR,βS)-rel-α-tert-Butyl-β-(4-chlorophenoxy)-1H-1,2,4-triazole-1-ethanol
Empirical Formula (Hill Notation):
C14H18ClN3O2
CAS Number:
Molecular Weight:
295.76
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing