
Terbuthylazin-desethyl

Synonym(s):
2-Amino-4-tert-butylamino-6-chloro-1,3,5-triazine, Desethylterbutylazine
Empirical Formula (Hill Notation):
C7H12ClN5
CAS Number:
Molecular Weight:
201.66
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing