Lovastatin
Synonym(s):
Lovastatin, Mevinolin
Empirical Formula (Hill Notation):
C24H36O5
CAS Number:
Molecular Weight:
404.54
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3