Fluoxetine Hydrochloride
Synonym(s):
Fluoxetine hydrochloride, (±)-N-Methyl-γ-[4-(trifluoromethyl)phenoxy]benzenepropanamine hydrochloride, LY-110,140 hydrochloride, Prozac®
Empirical Formula (Hill Notation):
C17H18F3NO · HCl
CAS Number:
Molecular Weight:
345.79
Products
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Product Line
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing