Skip to Content
Merck
TWEEN® 80 solution 10%, low peroxide

TWEEN® 80

TWEEN® 80 solution 10%, low peroxide
Synonym(s):
TWEEN® 80, POE (20) sorbitan monooleate, Polyethylene glycol sorbitan monooleate, Polyoxyethylenesorbitan monooleate, Polysorbate 80, Polyethylene glycol sorbitan monooleate solution, Polyoxyethylenesorbitan Monooleate
CAS Number:

Products

Product Category
Brand
Color
Feature
Physical Form
Quality Segment