Skip to Content
Merck
Sodium 3,3′-{[(1,2-diphenylethene-1,2-diyl)bis(4,1-phenylene)]bis(oxy)}bis(propane-1-sulfonate)

Sodium 3,3′-{[(1,2-diphenylethene-1,2-diyl)bis(4,1-phenylene)]bis(oxy)}bis(propane-1-sulfonate)

Sodium 3,3′-{[(1,2-diphenylethene-1,2-diyl)bis(4,1-phenylene)]bis(oxy)}bis(propane-1-sulfonate)
Empirical Formula (Hill Notation):
C32H30Na2O8S2
CAS Number:
Molecular Weight:
652.69

Products

Brand
Formula Weight
Physical Form
Showing 1-1 of 1