(S)-Naproxen ethyl ester
Synonym(s):
(2S)-2-(6-Methoxy-2-naphthyl)propionic acid ethyl ester, Ethyl (αS)-6-methoxy-α-methyl-2-naphthaleneacetate, Ethyl (2S)-2-(6-methoxy-2-naphthyl)propionate
Empirical Formula (Hill Notation):
C16H18O3
CAS Number:
Molecular Weight:
258.31
Products
Brand
Formula Weight
Purity
Format
Quality Segment
Application
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing