(R)-(−)-2-Phenylpropionic acid
Synonym(s):
(R)-(−)-Hydratropic acid, (R)-HTA
Linear Formula:
CH3CH(C6H5)CO2H
CAS Number:
Molecular Weight:
150.17
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing