
Punicalin, mixture of anomers

Synonym(s):
4,6-Gallagyl-D-glucose
Empirical Formula (Hill Notation):
C34H22O22
CAS Number:
Molecular Weight:
782.53
Products
Brand
Formula Weight
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing