Skip to Content
Merck
PCB No 153 solution 10 μg/mL in isooctane, analytical standard

PCB No 153 solution

PCB No 153 solution 10 μg/mL in isooctane, analytical standard
Synonym(s):
2,2′,4,4′,5,5′-Hexachlorobiphenyl solution
Empirical Formula (Hill Notation):
C12H4Cl6
CAS Number:
Molecular Weight:
360.88
EC Number:

Products

Brand
Formula Weight
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing