
Niobium diselenide

Synonym(s):
Niobium selenide
Empirical Formula (Hill Notation):
NbSe2
CAS Number:
Molecular Weight:
250.83
Products
Brand
Formula Weight
Physical Form
Purity
Height (mm)
Length (cm)
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing