Metrifonate
Synonym(s):
Trichlorfon, (2,2,2-Trichloro-1-hydroxyethyl)phosphonic acid dimethyl ester, Metrifonate
Empirical Formula (Hill Notation):
C4H8Cl3O4P
CAS Number:
Molecular Weight:
257.44
Products
Formula Weight
Manufacturer
Format
Application
API Family
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing