Skip to Content
Merck
Indeno[1,2,3-c,d]pyrene solution 100 μg/mL in cyclohexane, analytical standard

Indeno[1,2,3-c,d]pyrene solution

Indeno[1,2,3-c,d]pyrene solution 100 μg/mL in cyclohexane, analytical standard
Empirical Formula (Hill Notation):
C22H12
CAS Number:
Molecular Weight:
276.33

Products

Brand
Formula Weight
Format
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1