
Ethylbenzene-d10

Synonym(s):
Decadeuteroethylbenzene
Linear Formula:
C6D5CD2CD3
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Isotope
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing