
Ethyl 2-(trimethylsilylmethyl)acrylate

Synonym(s):
2-(Trimethylsilylmethyl)acrylic acid ethyl ester, Ethyl 2-(trimethylsilylmethyl)propenoate
Linear Formula:
H2C=C[CH2Si(CH3)3]CO2C2H5
CAS Number:
Molecular Weight:
186.32
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Monomer Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing