
Diamorphine hydrochloride

Synonym(s):
Heroin hydrochloride
Empirical Formula (Hill Notation):
C21H23NO5 · HCl
CAS Number:
Molecular Weight:
405.87
Products
Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing