
(+)-Fenchol

Synonym(s):
(1R)-1,3,3-Trimethylbicyclo[2.2.1]heptan-2-ol, Fenchyl alcohol
Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
Products
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Format
Quality Segment
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing