Dequalinium chloride for performance test
Synonym(s):
Dequalinium chloride, 1,1′-Decamethylenebis(4-aminoquinaldinium) dichloride
Empirical Formula (Hill Notation):
C30H40Cl2N4
CAS Number:
Molecular Weight:
527.57
Products
Formula Weight
Manufacturer
Melting Point (Ā°C)
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing