(−)-Corey lactone, 4-phenylbenzoate alcohol
Synonym(s):
(3aα,4α,5β,6aα)-(−)-Hexahydro-4-(hydroxymethyl)-2-oxo-2H-cyclopenta[b]furan-5-yl 1,1′-biphenyl-4-carboxylate, (3aR,4S,5R,6aS)-Hexahydro-4-hydroxymethyl-5-(4-phenylbenzoyloxy)cyclopenta[b]furan-2-one
Empirical Formula (Hill Notation):
C21H20O5
CAS Number:
Molecular Weight:
352.38
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing