
Ciprofibrate for system suitability

Synonym(s):
Ciprofibrate, 2-[p-(2,2-Dichlorocyclopropyl)phenoxy]-2-methylpropanoic acid
Empirical Formula (Hill Notation):
C13H14Cl2O3
CAS Number:
Molecular Weight:
289.15
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing