Chlortalidone for peak identification
Synonym(s):
Chlorthalidone
Empirical Formula (Hill Notation):
C14H11ClN2O4S
CAS Number:
Molecular Weight:
338.77
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing