CdSeS/ZnS alloyed quantum dots
Synonym(s):
Fluorescent Nanocrystals, Trilite™
Linear Formula:
CdSxSe1-x/ZnS
Products
Product Category
Brand
Boiling Point (°C)
Physical Form
Chemical Composition
Quality Segment
Functional Group
Showing 1-4 of 4
Compare
Product Number
Description
Pricing