Amobarbital sodium
Synonym(s):
Amobarbital sodium salt, 5-Ethyl-5-isoamylbarbituric acid, Amytal
Linear Formula:
C11H17N2O3Na
CAS Number:
Molecular Weight:
248.25
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing