Skip to Content
Merck
6-Nitro-2,3-dihydroxyquinoxaline 96%

6-Nitro-2,3-dihydroxyquinoxaline

6-Nitro-2,3-dihydroxyquinoxaline 96%
Synonym(s):
6-Nitroquinoxaline-2,3-dione
Empirical Formula (Hill Notation):
C8H5N3O4
CAS Number:
Molecular Weight:
207.14

Products

Product Category
Brand
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing